Learn More
Heidolph instruments Clamping Systems
Unique Pt 1000 clamping system that includes Clamping System Heidolph Pt1000, support rod, and attachment with cable guide that is ideal in research, scientific, and laboratory environments.
Brand: Heidolph instruments 509-63100-00
979.24 DKK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Pt 1000 clamping system
- Includes Clamping System Heidolph Pt1000, support rod, and attachment with cable guide
Order Info
dimProductType
Specifications
Clamping System |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
42Hours
21Minutes
31Seconds
38