Learn More
BRAND™ Class A Borosilicate Volumetric Flask with Polypropylene Stopper
Borosilicate volumetric flask with polypropylene stopper and ISO batch certificate.
Brand: BRAND™ 37233
325.81 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Borosilicate 3.3
- DIN EN ISO 1042
- Calibrated to contain (TC, In)
- All BLAUBRAND™ volumetric flasks are delivered with a batch number and batch certificate included in the original packaging. They are also available with individual certificates or DAkkS calibration certificate upon request.
Specifications
ISO Batch | |
Borosilicate Glass (3.3) | |
Reusable | |
Narrow | |
Flat |
150 mL | |
Autoclavable | |
Yes | |
Non-sterile |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Videos
-
Brand BLAUBRAND
Testing Instructions: BLAUBRAND Volumetric Instruments and Density Bottles
File size: 836.4 kB
BLAUBRAND Volumetric Instruments
File size: 896.8 kB
Brand Volumetric measurement in the laboratory
File size: 3.6 MB
Technical Note: Volumetric Flasks
File size: 194.3 kB
Special Offers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
New Offer Save Now
Product Content Correction
Product Title
BRAND™ Class A Borosilicate Volumetric Flask with Polypropylene Stopper > Volumetric Flask, 150 mL
Spot an opportunity for improvement?Share a Content Correction