Learn More
BRAND™ Class A Borosilicate Volumetric Flask with Polypropylene Stopper
Borosilicate volumetric flask with polypropylene stopper and ISO batch certificate.
435.00 DKK
Specifications
Certifications/Compliance | ISO Batch |
---|---|
Material | Borosilicate Glass (3.3) |
Autoclavable | Autoclavable |
Disposable | Reusable |
Graduated | Yes |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
16445989
|
BRAND™
37233 |
150 mL |
435.00 DKK
Pack of 2 |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
Description
- Borosilicate 3.3
- DIN EN ISO 1042
- Calibrated to contain (TC, In)
- All BLAUBRAND™ volumetric flasks are delivered with a batch number and batch certificate included in the original packaging. They are also available with individual certificates or DAkkS calibration certificate upon request.
Specifications
ISO Batch | |
Autoclavable | |
Yes | |
Flat |
Borosilicate Glass (3.3) | |
Reusable | |
Non-sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BRAND™ Class A Borosilicate Volumetric Flask with Polypropylene Stopper