Learn More
Thermo Scientific™ ClipTip™ Pipette Tip Racks
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
Brand: Thermo Scientific™ 94410219
167.10 DKK valid until 2025-06-30
Use promo code "25076" to get your promotional price.
Description
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
- Small Rack: For use with ClipTip 20 and 50 Pipette Tips
- Medium Rack: For use with ClipTip 200 and 300 Pipette Tips
- Large Rack: For use with ClipTip 1000 Pipette Tips
Ordering Information:
Racks are supplied without tips and inserts
Specifications
ClipTip Empty Rack, Small | |
ClipTip Empty Rack | |
Thermo Scientific E1-ClipTip | |
ClipTip |
Special Offers
-
Thermo Scientific ClipTip
,
Thermo Scientific ART
,
Thermo Scientific Finnpipette
,
Thermo Scientific Matrix
Exclusive Promotion on Liquid Handling
Unlock Precision: Top Pipettes and Tips on Promotion!Promo Code: 25076Expires: 2025-06-30