Learn More
Thermo Scientific™ ClipTip™ Pipette Tip Racks
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
248.00 DKK - 251.00 DKK
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
13256249
![]() |
Thermo Scientific™
94410219 |
ClipTip Empty Rack, Small |
251.00 DKK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
15922021
![]() |
Thermo Scientific™
94410619 |
ClipTip Empty Rack |
250.00 DKK
Pack of 8 |
In Stock
Log in to see stock. |
||||||
13286259
![]() |
Thermo Scientific™
94410519 |
ClipTip Empty Rack, Medium |
250.00 DKK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
13236269
![]() |
Thermo Scientific™
94410819 |
ClipTip Empty Rack, Large |
248.00 DKK
Pack of 8 |
In Stock
Log in to see stock. |
||||||
Description
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
- Small Rack: For use with ClipTip 20 and 50 Pipette Tips
- Medium Rack: For use with ClipTip 200 and 300 Pipette Tips
- Large Rack: For use with ClipTip 1000 Pipette Tips
Ordering Information:
Racks are supplied without tips and inserts
Specifications
ClipTip Empty Rack | |
ClipTip |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Thermo Scientific™ ClipTip™ Pipette Tip Racks