Learn More
WTW™ Connection Cable
Brand: WTW™ 903850
731.17 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
MPP IDS Respectively IDS Sensors | |
Connection Cable | |
1.5 m |
Product Content Correction
Product Title
WTW™ Connection Cable > Length: 1.5m
Spot an opportunity for improvement?Share a Content Correction