missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated CD68 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-25747-20UL
This item is not returnable.
View return policy
Description
The CD68 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human CD68. This antibody recognizes human, mouse antigen. The CD68 antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.Specifications
CD68 | |
Polyclonal | |
CoraLite Plus 488 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
968 | |
Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
Conjugated Antibody | |
IgG |
Immunofluorescence | |
1000 ÎĽg/mL | |
DKFZp686M18236; GP110; SCARD1 | |
Fusion Protein | |
20 ÎĽL | |
Primary | |
Human, Mouse | |
60 kDa | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CoraLite™ Plus 488-conjugated CD68 Rabbit Polyclonal Antibody, Proteintech > CoraLite™ Plus 488; 20 μL
Spot an opportunity for improvement?Share a Content Correction