missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated CD68 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
968.42 DKK - 2619.89 DKK
Specifications
Antigen | CD68 |
---|---|
Concentration | 1000 ÎĽg/mL |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | CoraLite Plus 488 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30235672
|
Proteintech
CL488-25747-100UL |
100 ÎĽL |
2619.89 DKK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30236260
|
Proteintech
CL488-25747-20UL |
20 ÎĽL |
968.42 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD68 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human CD68. This antibody recognizes human, mouse antigen. The CD68 antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.Specifications
CD68 | |
Immunofluorescence | |
CoraLite Plus 488 | |
Rabbit | |
Human, Mouse | |
DKFZp686M18236; GP110; SCARD1 | |
IgG | |
Antigen affinity purification | |
Conjugated Antibody |
1000 ÎĽg/mL | |
Polyclonal | |
Liquid | |
RUO | |
968 | |
Fusion Protein | |
Primary | |
Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CoraLite™ Plus 488-conjugated CD68 Rabbit Polyclonal Antibody, Proteintech