Learn More
Corning™ Costar™ Thermowell™ Plate Lid
Fits styles C, M, and H.
Brand: Corning™ 6520
918.35 DKK valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description
- Polycarbonate lids are thermally stable up to 135°C and can be used on Corning™ Thermowell™ 96 well polycarbonate plates (fits Cat. No. 6511) with oil overlays
- Lids are intended to be covers only and do not prevent evaporation
Specifications
Polycarbonate | |
96 | |
1/Pk. | |
Thermowell Polycarbonate Plates |
Non-sterile | |
25/Cs. | |
Plate Lid |
Product Suggestions
Customers who viewed this item also viewed
COR001 Lifescience Guid
File size: 7 MB
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Product Content Correction
Product Title
Corning™ Costar™ Thermowell™ Plate Lid > Polycarbonate; 25/Pk; 96-well
Spot an opportunity for improvement?Share a Content Correction