Learn More
Dabigatran-d4 hydrochloride, MedChemExpress
MedChemExpress Dabigatran (BIBR-953) D4 hydrochloride is deuterium labeled Dabigatran, which is a reversible and selective, direct thrombin inhibitor (DTI) with a Ki value of 4.5 nM.
Brand: Medchem Express HY-10163AS
This item is not returnable.
View return policy
Description
Dabigatran (BIBR-953) D4 hydrochloride is deuterium labeled Dabigatran, which is a reversible and selective, direct thrombin inhibitor (DTI) with a Ki value of 4.5 nM.Specifications
Dabigatran-d4 hydrochloride | |
Colorless | |
C25H22D4ClN7O3 | |
Neuroscience-Neuromodulation | |
N=C(N)C(C([2H])=C1[2H])=C([2H])C([2H])=C1NCC2=NC(C=C3C(N(CCC(O)=O)C4=NC=CC=C4)=O)=C(C=C3)N2C.Cl | |
512.0 | |
Research |
5 mg | |
Research | |
BIBR-953-d4 hydrochloride | |
Solid | |
512.0 | |
97.05% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Dabigatran-d4 hydrochloride, MedChemExpress