missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol, Extra Pure, SLR, Fisher Chemical™

245.00 DKK - 4480.00 DKK

Chemical Identifiers

CAS 111-46-6
Molecular Formula C4H10O3
Molecular Weight (g/mol) 106.12
MDL Number MFCD00002882
InChI Key MTHSVFCYNBDYFN-UHFFFAOYSA-N
Synonym diethylene glycol, 2,2'-oxydiethanol, diglycol, diethylenglykol, 2-hydroxyethyl ether, bis 2-hydroxyethyl ether, ethanol, 2,2'-oxybis, 2,2'-oxybisethanol, 2-2-hydroxyethoxy ethanol, digol
PubChem CID 8117
ChEBI CHEBI:46807
SMILES OCCOCCO
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10428020
View Documents
Fisher Chemical
D/2950/08
500 mL Amber glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10438020
View Documents
Fisher Chemical
D/2950/17
2.5 L Amber glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12688625
View Documents
Fisher Chemical
D/2950/25
25 L Plastic container This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

111-46-6
106.12
MTHSVFCYNBDYFN-UHFFFAOYSA-N
8117
OCCOCCO
C4H10O3
MFCD00002882
diethylene glycol, 2,2'-oxydiethanol, diglycol, diethylenglykol, 2-hydroxyethyl ether, bis 2-hydroxyethyl ether, ethanol, 2,2'-oxybis, 2,2'-oxybisethanol, 2-2-hydroxyethoxy ethanol, digol
CHEBI:46807

Specifications

111-46-6
Colorless
C4H10O3
diethylene glycol, 2,2'-oxydiethanol, diglycol, diethylenglykol, 2-hydroxyethyl ether, bis 2-hydroxyethyl ether, ethanol, 2,2'-oxybis, 2,2'-oxybisethanol, 2-2-hydroxyethoxy ethanol, digol
OCCOCCO
106.12
CHEBI:46807
Liquid
-10°C
245°C
MFCD00002882
MTHSVFCYNBDYFN-UHFFFAOYSA-N
2-(2-hydroxyethoxy)ethan-1-ol
8117
106.12g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Diethylene glycol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H373-May cause damage to organs.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.