missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIS3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29664-1-AP-150UL
This item is not returnable.
View return policy
Description
DIS3 Polyclonal antibody apecifically detetects DIS3 antigen in Human, Mouse Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.Specifications
DIS3 | |
Polyclonal | |
Unconjugated | |
NM_014953 | |
Rabbit | |
Antigen affinity purification | |
Primary | |
Human, Mouse | |
Liquid |
Western Blot, Immunohistochemistry, ELISA | |
300 μg/mL | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
DIS3 | |
DIS3 fusion protein Ag31711 | |
150 μL | |
22894 | |
Store at -20°C. Stable for one year after shipment. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DIS3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction