missing translation for 'onlineSavingsMsg'
Learn More

DIS3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1081.84 DKK - 3058.99 DKK

Specifications

Antigen DIS3
Concentration 300 ÎĽg/mL
Applications Western Blot, Immunohistochemistry, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17689676
View Documents
Proteintech
29664-1-AP-20UL
20 ÎĽL
1081.84 DKK
20µL
Please to purchase this item. Need a web account? Register with us today!
17679676
View Documents
Proteintech
29664-1-AP-150UL
150 ÎĽL
3058.99 DKK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DIS3 Polyclonal antibody apecifically detetects DIS3 antigen in Human, Mouse Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.
Specifications

Specifications

DIS3
Western Blot, Immunohistochemistry, ELISA
Unconjugated
Rabbit
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
22894
DIS3 fusion protein Ag31711
Primary
Store at -20°C. Stable for one year after shipment.
300 ÎĽg/mL
Polyclonal
Liquid
Human, Mouse
NM_014953
DIS3
IgG
Antigen affinity purification
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DIS3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.