Learn More
DMP 777, MedChemExpress
MedChemExpress DMP 777 is a potent, selective, and orally active human leukocyte elastase (HLE) inhibitor.
Brand: Medchem Express HY-75957
This item is not returnable.
View return policy
Description
DMP 777 is a potent, selective, and orally active human leukocyte elastase (HLE) inhibitor.Specifications
DMP 777 | |
Off-White | |
C31H40N4O6 | |
L-694458 | |
CCC[C@H](C1=CC2=C(C=C1)OCO2)NC(N3[C@H](C(CC)(C3=O)CC)OC4=CC=C(C=C4)C(N5CCN(CC5)C)=O)=O | |
564.67 | |
Research | |
COVID-19-immunoregulation |
157341-41-8 | |
Research | |
25 mg | |
DMSO : 38.33 mg/mL (67.88 mM; Need ultrasonic) | |
564.67 | |
98.41% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.