Learn More
Eldecalcitol-d6, MedChemExpress
MedChemExpress Eldecalcitol-d6 is the deuterium labeled Eldecalcitol. Eldecalcitol is an orally active analogue of active vitamin D used in the treatment of osteoporosis.
Brand:  Medchem Express HY-A0020S
Encompass_Preferred
Product Code. 16402009
  • 28716.28 DKK / 1mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
Eldecalcitol-d6 is the deuterium labeled Eldecalcitol. Eldecalcitol is an orally active analogue of active vitamin D used in the treatment of osteoporosis.
Specifications
Specifications
Eldecalcitol-d6
White
C30H44D6O5
C=C1[C@@H](O)[C@H](OCCCO)[C@H](O)C/C1=C/C=C2CCC[C@@]3(C)[C@@]\2([H])CC[C@@H]3[C@@H](CCCC(O)(C([2H])([2H])[2H])C([2H])([2H])[2H])C
496.75
Research
Solid
1 mg
Research
DMSO : 25 mg/mL (50.33 mM; Need ultrasonic)
496.75
99.26%
Neuroscience-Neuromodulation