Learn More
Entacapone, MedChemExpress
MedChemExpress Entacapone is a potent, reversible, peripherally acting and orally active catechol-O-methyltransferase (COMT) inhibitor.
Brand:  Medchem Express HY-14280
Encompass_Preferred
 View more versions of this product
Product Code. 16406436
  • 1873.25 DKK / 100mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
Entacapone inhibits COMT from rat brain, erythrocytes and liver with IC50 values of 10 nM, 20 nM, and 160 nM, respectively. Entacapone is selective for COMT over other catecholamine metabolizing enzymes, including MAO-A, MAO-B, phenolsulphotransferase M (PST-M) and PST-P (IC50s>50 μM). Entacapone can be used for the research of Parkinson's disease. Entacapone serves as a inhibitor of FTO demethylation with an IC50 of 3.5 μM, can be used for the research of metabolic disorders.
Specifications
Specifications
Entacapone
Yellow
C14H15N3O5
DMSO : 33.33 mg/mL (109.17 mM; Need ultrasonic) ∣H2O : 2 mg/mL (6.55 mM; ultrasonic and adjust pH to 10 with NaOH)
305.29
98.79%
Powder
130929-57-6
Research
100 mg
O=C(N(CC)CC)/C(C#N)=C/C1=CC([N+]([O-])=O)=C(O)C(O)=C1
305.29
Research
Neuroscience-Neurodegeneration
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Entacapone, MedChemExpress