missing translation for 'onlineSavingsMsg'
Learn More

National Lab Eppi™45 Kryo Boxes

Brand:  National Lab KEA45-V81NO

Additional Details : Weight : 0.30000kg

 View more versions of this product

Product Code. 12444800

  • 33.00 DKK / Each
Estimated Shipment: 19-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Neon Orange
81 Vials up to 12.5 mm Diameter
130 x 130 mm
130 mm
Rectangular
Kryo Box
130 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers