missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol Absolute, Solveco
Fermented and rectified ethanol of very high purity, suitable for routine laboratory and analysis. The alcohol content reaches 99.9%.
660.00 DKK - 672.00 DKK
Chemical Identifiers
CAS | 64-17-5 |
---|---|
Molecular Formula | C2H6O |
Molecular Weight (g/mol) | 46.069 |
InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
Synonym | ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol |
PubChem CID | 702 |
ChEBI | CHEBI:16236 |
IUPAC Name | ethanol |
SMILES | CCO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11594139
|
Solveco Chemicals
1015 |
12 x 1 L |
660.00 DKK
Pack of 12 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16661932
|
Solveco Chemicals
1395 |
3 x 5 L |
672.00 DKK
Pack of 3 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Well suited for routine laboratory and analysis. Solveco Ethanol Absolute is fermented and rectified ethanol with very high purity. This PA quality solution has a natural origin.
Chemical Identifiers
64-17-5 | |
46.069 | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsolShow More | |
CHEBI:16236 | |
CCO |
C2H6O | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
702 | |
ethanol |
Specifications
≤0.500 (UV 5 cm, 240 nm) | |
Colorless | |
78°C | |
C2H6O | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
ethanol | |
702 | |
46.07g/mol | |
AG |
64-17-5 | |
≤0.790 g/cm3 | |
HDPE Bottle | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CCO | |
46.069 | |
CHEBI:16236 | |
99.9% | |
Ethyl Alcohol |
Safety and Handling
Recommended Storage : Store at 18°C to 30°C for up to 2 years in sealed package.
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Etanol 99,5%
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ethanol Absolute, Solveco