Learn More
Memmert™ Extended Overtemperature Protection for SF260
Extended overtemperature protection with integrated Pt100 sensor for independent temperature monitoring.
Brand:  Memmert™ SF260A6
Product Code. 17558165
  • 1425.00 DKK / Each
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Description
  • Accessory
  • With integrated Pt100 sensor
  • For models with SingleDISPLAY
Specifications
Specifications
SF260 Sterilizer
Integrated Pt100 Sensor
Product Title
Memmert™ Extended Overtemperature Protection for SF260 > For Use With: SF260