Learn More
Fibronectin Mouse anti-Human, Bovine, Chicken, Clone: A17, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Bio-Techne NBP2-23524-0.025MG
 View more versions of this product
Product Code. 18197732
  • 1545.00 DKK / 25µg
Explore more special offers
This item is not returnable. View return policy
Description
Description
Fibronectin Monoclonal specifically detects Fibronectin in Human, Bovine, Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fibronectin
Monoclonal
1 mg/ml
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500
CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF
Lysed bovine corneal endothelial cells and extracellular matrix.
0.025 mg
Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Stem Cell Markers
2335
Store at 4°C. Do not freeze.
IgG1
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation
A17
Unconjugated
10mM Phosphate (pH 7.4) and 0.5M NaCl
Mouse
Protein A or G purified
RUO
Primary
Human, Bovine, Chicken
Purified