missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibronectin Mouse anti-Human, Bovine, Chicken, Clone: A17, Novus Biologicals™
Mouse Monoclonal Antibody
1545.00 DKK - 3775.00 DKK
Specifications
Antigen | Fibronectin |
---|---|
Clone | A17 |
Concentration | 1 mg/ml |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500 |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18431512
|
Novus Biologicals
NBP2-23524-100ug |
100 ug |
3775.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18197732
|
Novus Biologicals
NBP2-23524-0.025MG |
0.025 mg |
1545.00 DKK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fibronectin Monoclonal specifically detects Fibronectin in Human, Bovine, Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Fibronectin | |
1 mg/ml | |
Monoclonal | |
Purified | |
RUO | |
10mM Phosphate (pH 7.4) and 0.5M NaCl | |
2335 | |
IgG1 | |
Protein A or G purified |
A17 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500 | |
Unconjugated | |
Mouse | |
Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
Lysed bovine corneal endothelial cells and extracellular matrix. | |
Primary | |
Store at 4°C. Do not freeze. |