missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBPL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10060-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene has similarity to the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. The encoded protein is thought to have a potential role in the induced radioresistance. Also it appears to have some involvement in the control of the cell cycle.Specifications
FKBPL | |
Polyclonal | |
Unconjugated | |
FKBPL | |
DIR1, FK506 binding protein like, FKBPL, NG7, WISP39 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
406168, 56299, 63943 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35450, Q6MG81, Q9UIM3 | |
Fkbpl | |
FKBPL Fusion Protein Ag0112 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FKBPL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction