missing translation for 'onlineSavingsMsg'
Learn More

FKBPL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1081.84 DKK - 3058.99 DKK

Specifications

Antigen FKBPL
Concentration 0.26 mg/mL
Applications Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16886203
View Documents
Proteintech
10060-1-AP-20UL
20 ÎĽL
1081.84 DKK
20µL
Please to purchase this item. Need a web account? Register with us today!
16876203
View Documents
Proteintech
10060-1-AP-150UL
150 ÎĽL
3058.99 DKK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The protein encoded by this gene has similarity to the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. The encoded protein is thought to have a potential role in the induced radioresistance. Also it appears to have some involvement in the control of the cell cycle.
Specifications

Specifications

FKBPL
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Mouse, Human, Rat
O35450, Q6MG81, Q9UIM3
406168, 56299, 63943
FKBPL Fusion Protein Ag0112
Primary
-20°C
FKBPL
0.26 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
DIR1, FK506 binding protein like, FKBPL, NG7, WISP39
Fkbpl
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
FKBPL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.