Learn More
Lenz Laborglasinstrumente™ Flat Flange Lid with Center Neck, Three Angled Side Necks, and One Vertical Side Neck
Brand: Lenz Laborglasinstrumente™ 06105264
Specifications
Borosilicate Glass, Duran Glass | |
Laboratory Flange 100 | |
Center Neck (NS): 45/40 |
Reaction vessels | |
Flange lid |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.