missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FLJ20186 Recombinant Protein
Specifications
Accession Number | AAH15482 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 54849 |
Molecular Weight (g/mol) | 47.41 |
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH15482 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
47.41 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DEF8 | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
54849 | |
FLJ20186 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEYDEKLARFRQAHLNPFNKQSGPRQHEQGPGEEVPDVTPEEALPELPPGEPEFRCPERVMDLGLSEDHFSRPVGLFLASDVQQLRQAIEECKQVILELPEQSEKQKDAVVRLIHLRLKLQELKDPNEDEPNIRVLLEHRFYKEKSKSVKQTCDKCNTIIWGLIQTWYTCTGGPRPRRGVRNERDQSSCLRWAHIQM | |
FLJ20186/MGC104349 | |
DEF8 | |
Wheat Germ (in vitro) | |
GST |