Learn More
Euro Protection™ Freedite Low-Cut Safety Shoes
Euro Protection™ Freedite Low-Cut Safety Shoes are padded for a comfortable, snug fit and feature a footbridge for rigidity and a composite toe cap for safety.
466.00 DKK - 517.00 DKK
Specifications
Color | Black |
---|---|
Liner Material | Breathable 3D mesh lining |
Material | Leather |
Sole Features | Removable, textile and foam insole, non-metallic (composite fabric) puncture-resistant sole, injected polyurethane midsole, polyurethane outsole. |
Sole Material | Polyurethane, Composite Fabric |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
15795625
|
Euro Protection™
9FREL36 |
36 |
466.00 DKK
1 pair |
Estimated Shipment: 10-04-2025
Log in to see stock. |
||||||
15705635
|
Euro Protection™
9FREL37 |
37 |
466.00 DKK
1 pair |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||||
15715635
|
Euro Protection™
9FREL38 |
38 |
466.00 DKK
1 pair |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||||
15725635
|
Euro Protection™
9FREL39 |
39 |
466.00 DKK
1 pair |
Estimated Shipment: 10-04-2025
Log in to see stock. |
||||||
15735635
|
Euro Protection™
9FREL40 |
40 |
504.00 DKK
1 pair |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||||
15745635
|
Euro Protection™
9FREL41 |
41 |
504.00 DKK
1 pair |
Estimated Shipment: 10-04-2025
Log in to see stock. |
||||||
15755635
|
Euro Protection™
9FREL42 |
42 |
504.00 DKK
1 pair |
Estimated Shipment: 10-04-2025
Log in to see stock. |
||||||
15765635
|
Euro Protection™
9FREL43 |
43 |
517.00 DKK
1 pair |
Estimated Shipment: 19-03-2025
Log in to see stock. |
||||||
15775635
|
Euro Protection™
|
44 |
N/A
|
N/A | ||||||
15785635
|
Euro Protection™
9FREL45 |
45 |
504.00 DKK
1 pair |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||||
15795635
|
Euro Protection™
9FREL46 |
46 |
466.00 DKK
1 pair |
Estimated Shipment: 04-04-2025
Log in to see stock. |
||||||
15705645
|
Euro Protection™
9FREL47 |
47 |
466.00 DKK
1 pair |
Estimated Shipment: 04-04-2025
Log in to see stock. |
||||||
15715645
|
Euro Protection™
9FREL48 |
48 |
466.00 DKK
1 pair |
Estimated Shipment: 04-04-2025
Log in to see stock. |
||||||
Description
- Footbridge for torsional rigidity.
- Padding for snug fit.
- Also available with an electrical hazards resistant outsole on request (Reference: 9FEHH).
- Full grain leather upper, reflective trim at the back.
- Breathable 3D mesh lining.
- Removable, textile and foam insole.
- Composite toe cap.
- Non-metallic (composite fabric) puncture resistant sole.
- Injected polyurethane midsole.
- Polyurethane outsole.
Specifications
Black | |
Leather | |
Polyurethane, Composite Fabric | |
Composite | |
EN ISO 20345 : 2011 |
Breathable 3D mesh lining | |
Removable, textile and foam insole, non-metallic (composite fabric) puncture-resistant sole, injected polyurethane midsole, polyurethane outsole. | |
Low | |
563 g per shoe in size 8 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Euro Protection™ Freedite Low-Cut Safety Shoes