Learn More
BRAND™ Gay-Lussac Uncalibrated Density Bottles
Gay-Lussac density bottle made with Borosilicate Glass 3.3 that is ideal in most research, scientific, and other laboratory environments.
Brand:  BRAND™ 43208
519.28 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11808291
  • 662.00 DKK / Pack of 2
Estimated Shipment: 28-05-2025
Explore more special offers
Description
  • BRAND™ density bottle
  • Gay-Lussac pattern
  • Uncalibrated capacity, 10 mL
  • Borosilicate glass, bottle narrow-mouth, bottle round (Gay-Lussac), white bottle
  • DIN ISO 3507
  • Stopper NS 10/19 with capillary
  • Top of stopper ground and polished
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2Each
10 mL
Borosilicate Glass (3.3)
Gay-Lussac Density Bottle
Product Title
BRAND™ Gay-Lussac Uncalibrated Density Bottles > Volume: 10mL