Learn More
BRAND™ Gay-Lussac Uncalibrated Density Bottles
Gay-Lussac density bottle made with Borosilicate Glass 3.3 that is ideal in most research, scientific, and other laboratory environments.
Brand: BRAND™ 43228
618.51 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- BRAND™ density bottle
- Gay-Lussac pattern
- Uncalibrated capacity, 10 mL
- Borosilicate glass, bottle narrow-mouth, bottle round (Gay-Lussac), white bottle
- DIN ISO 3507
- Stopper NS 10/19 with capillary
- Top of stopper ground and polished
Specifications
2Each | |
50 mL | |
Borosilicate Glass (3.3) | |
Gay-Lussac Density Bottle |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
BRAND™ Gay-Lussac Uncalibrated Density Bottles > Volume: 50mL
Spot an opportunity for improvement?Share a Content Correction