Learn More
Gilmont™ Gilmont™ Micrometer Syringes and Burets
Experience high precision with economy of cost when using Gilmont™ Micrometer Syringes.
Brand:  Gilmont™ 15-078-150
Encompass
 View more versions of this product
Product Code. 12367279
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
Experience high precision with economy of cost when using Thermo Scientific™ Gilmont Micrometer Syringes.
  • 0.5% accuracy
  • Liquid comes in contact with only glass, VITON™ and PTFE™
  • VITON O-ring gives a vacuum tight seal
  • All other plastic parts of polypropylene--instrument may be autoclaved
  • Available in two sizes, 0.2 mL in 0.0002 mL div. and 2.0 mL in 0.002 mL div.
  • Fills and delivers liquid with micrometer control
  • Simple to clean and assemble
  • Specifications
    Specifications
    Syringe
    0.2 mL
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.