Learn More
Gilmont™ Gilmont™ Micrometer Syringes and Burets
Experience high precision with economy of cost when using Gilmont™ Micrometer Syringes.
POA
Products 1
Product Code Brand Volume (Metric) Product Type Price Quantity & Availability  
Product Code Brand Volume (Metric) Product Type Price Quantity & Availability  
12367279
Encompass
View Documents
Gilmont™
15-078-150
0.2 mL Syringe This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Experience high precision with economy of cost when using Thermo Scientific™ Gilmont Micrometer Syringes.
  • 0.5% accuracy
  • Liquid comes in contact with only glass, VITON™ and PTFE™
  • VITON O-ring gives a vacuum tight seal
  • All other plastic parts of polypropylene--instrument may be autoclaved
  • Available in two sizes, 0.2 mL in 0.0002 mL div. and 2.0 mL in 0.002 mL div.
  • Fills and delivers liquid with micrometer control
  • Simple to clean and assemble
  • Videos
    SDS
    Documents
    Documents
    Product Certifications
    Product Content Correction
    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.