missing translation for 'onlineSavingsMsg'
Learn More

Woodsafe™ Sharpsbin and Hazardous waste containers with single lid in natural color
GREENER_CHOICE

Woodsafe is a cost-effective solution for reducing the use of fossil based plastic by 80% and CO2 emissions by 66%, documented in in a full LCA. Produced with residue from the forest industry using green energy. Available in sizes from 0.5 L to 50 L. Single and double lid versions.

Brand:  Woodsafe™ 1250110

121.98 DKK valid until 2025-05-30
Use promo code "24004" to get your promotional price.


 View more versions of this product

Product Code. 18196539

  • 128.40 DKK / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Alert:

Follow instructions for safe and correct use


Description

Description

  • Woodsafe™ offer a sustainable clinical waste management solution produced with residue from the Swedish forest industry
  • Lowers the use of Fossil-based plastic by 80%!
  • Lowers the carbon footprint by more than 60%, proved in full LCA
  • Made from 100% Biocomposite
  • The lid can be securely permanently closed for transportation to the final disposal
  • My Green Labs ACT Label
  • Follows same process as yellow bins and can be used to transport the same substances
  • Natural color and smell from wood - No migration of substances
  • Compliant with UN3291
  • Tested according to ISO 23907-1:2019
  • UN ADR Approved
  • Biocomposite have the same properties as fossil-based plastic and can be used exactly the same way
  • NOTE: String between lids is for delivery purpose, remove/break before use!
  • Stop burning plastic
Specifications

Specifications

25 L
Permanent closure
Environmental friendly Sharpsbin
Brown, Yellow
Single-use
Non-sterile
Bin, Lid, and Label
Pallet
Biohazard Waste Container
EN ISO 23907-1:2019. UN3291, ADR Approved
Brown
6.157 gal
39.5 cm x 39.5 cm x 23.8 cm (15.6 x 15.6 x 9.4 in.)
Yes
Autoclavable
Safe Disposal of sharps and potentially Infectious Waste or medical waste
Biocomposite
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Woodsafe™ Sharpsbin and Hazardous waste containers with single lid in natural color > Capacity: 25 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Lower climate impact and ACT label