missing translation for 'onlineSavingsMsg'
Learn More

Histamine H4R Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Bio-Techne NBP3-03549-100UL

 View more versions of this product

Product Code. 18310906

  • 3498.83 DKK / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Histamine H4R Polyclonal specifically detects Histamine H4R in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

Histamine H4R
Polyclonal
PBS with 50% glycerol, pH7.3.
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
Unconjugated
AXOR35H4, GPCR105MGC133027, G-protein coupled receptor 105, GPRv53histamine H4 receptor, H4Rpfi-013, HH4RBG26, histamine receptor H4, Pfi-013, SP9144
Recombinant fusion protein containing a sequence corresponding to amino acids 194-304 of human Histamine H4R (NP_067637.2).
100 μL
GPCR, Immunology, Innate Immunity
59340
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers