missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histamine H4R Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1485.92 DKK - 3498.83 DKK
Specifications
Antigen | Histamine H4R |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18345776
|
Bio-Techne
NBP3-03549-20UL |
20 μL |
1485.92 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18310906
|
Bio-Techne
NBP3-03549-100UL |
100 μL |
3498.83 DKK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histamine H4R Polyclonal specifically detects Histamine H4R in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Histamine H4R | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
AXOR35H4, GPCR105MGC133027, G-protein coupled receptor 105, GPRv53histamine H4 receptor, H4Rpfi-013, HH4RBG26, histamine receptor H4, Pfi-013, SP9144 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 194-304 of human Histamine H4R (NP_067637.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
GPCR, Immunology, Innate Immunity | |
PBS with 50% glycerol, pH7.3. | |
59340 | |
IgG | |
Affinity purified |