Learn More
Jaytec Glass™ Hoffman Voltameter Glass Unit
The Hoffman Voltameter Glass Unit is for classroom demonstration of electrolysis to determine the chemical composition of water.
Brand:  Jaytec Glass™ EKY-710-L
 View more versions of this product
Product Code. 12391429
  • 1435.00 DKK / Each
Estimated Shipment: 12-08-2025
to see stock.
Explore more special offers
Description
  • Manufactured in borosilicate glass
  • Available Graduated or Ungraduated
  • H-pattern style
  • 50 mL capacity
Specifications
Specifications
Graduated Hoffman Voltameter
To demonstrate electrolysis of water into oxygen and hydrogen
0 to 50 mL x 0.2 mL
Documents
One moment while we fetch your results.
Certificates
View accessory products
Product Title
Jaytec Glass™ Hoffman Voltameter Glass Unit > Hoffman Voltameter Glass Unit; Type: Graduated; Capacity: 0 to 50 mL x 0.2 mL