Learn More
Jaytec Glass™ Hoffman Voltameter Glass Unit
The Hoffman Voltameter Glass Unit is for classroom demonstration of electrolysis to determine the chemical composition of water.
1110.00 DKK - 1435.00 DKK
Specifications
For Use With (Application) | To demonstrate electrolysis of water into oxygen and hydrogen |
---|---|
Quantity | 0 to 50 mL x 0.2 mL |
Product Code | Brand | Product Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | ||||||
12391429
|
Jaytec Glass™
EKY-710-L |
Graduated Hoffman Voltameter |
1435.00 DKK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||||
12381429
|
Jaytec Glass™
EKY-700-S |
Ungraduated Hoffman Voltameter |
1110.00 DKK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||||
Description
- Manufactured in borosilicate glass
- Available Graduated or Ungraduated
- H-pattern style
- 50 mL capacity
Specifications
To demonstrate electrolysis of water into oxygen and hydrogen | |
0 to 50 mL x 0.2 mL |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Jaytec Glass™ Hoffman Voltameter Glass Unit