Learn More
SCAT™ HPLC Air Valve, for PFAS Analytic
GREENER_CHOICE
The SCAT air valve blocks harmful vapours and ensures safe pressure equalisation in the storage tanks of your HPLC. Solvent mixtures remain stable: thanks to closed system without evaporation.
Brand:  SCAT™ 397008-PP
 View more versions of this product
Product Code. 18696488
  • 1795.00 DKK / 8 sets
Estimated Shipment: 25-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • With Luer-Lock connection for gassing the solvents and change indicator for timely change.
  • Made of PP for PFAS analysis of polyfluorinated chemicals (formerly: PFC)
  • UNF 1/4 in. 28G thread size fits to any SCA PFAS-Analytic Safety Cap for PFAS Analysis
Specifications
Specifications
Red, Green
SCAT Safety Caps for PFAS-Analytic
8 Pk.
Valve
HPLC analysis
PP
UNF 1/4 in. 28G
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SCAT™ HPLC Air Valve, for PFAS Analytic > Quantity: 8 Pk.