missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ HPLC Air Valve, for PFAS Analytic
GREENER_CHOICE

The SCAT air valve blocks harmful vapours and ensures safe pressure equalisation in the storage tanks of your HPLC. Solvent mixtures remain stable: thanks to closed system without evaporation.

POA - 2130.00 DKK

Specifications

Color Red, Green
Type Valve
For Use With (Application) HPLC analysis
For Use With (Equipment) SCAT Safety Caps for PFAS-Analytic
Material PP
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18686458
GREENER_CHOICE
View Documents
SCAT™
317011
1 Ea. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
18696488
GREENER_CHOICE
View Documents
SCAT™
397008-PP
8 Pk.
1795.00 DKK
8 sets
Estimated Shipment: 20-05-2025
Add to basket
 
18606498
GREENER_CHOICE
View Documents Promotion Details
SCAT™
397010-PP
10 Pk.
2130.00 DKK
10 sets
Estimated Shipment: 20-05-2025
Add to basket
 
Description

Description

  • With Luer-Lock connection for gassing the solvents and change indicator for timely change.
  • Made of PP for PFAS analysis of polyfluorinated chemicals (formerly: PFC)
  • UNF 1/4 in. 28G thread size fits to any SCA PFAS-Analytic Safety Cap for PFAS Analysis
Specifications

Specifications

Red, Green
HPLC analysis
PP
Valve
SCAT Safety Caps for PFAS-Analytic
UNF 1/4 in. 28G
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SCAT™ HPLC Air Valve, for PFAS Analytic

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.