Learn More
Invitrogen™ Human BRUCE Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP1454
Description
BRUCE Synthetic Peptide, PEP-1454, from Invitrogen.
Apoptosis, or programmed cell death, is related to many diseases, such as cancer. Apoptosis is triggered by a variety of stimuli including members in the TNF family and can be prevented by the inhibitor of apoptosis (IAP) proteins. IAP proteins form a conserved gene family that binds to and inhibits cell death proteases. BRUCE, also known as BIRC6, is an IAP family member protein with a BIR (baculoviral inhibition of apoptosis protein repeat) domain and a UBCc (ubiquitin-conjugating enzyme E2, catalytic) domain. BRUCE regulates p53 and the mitochondrial pathway of apoptosis by facilitating the degradation of apoptotic proteins such as Caspase-9 and SMAC by ubiquitination.
Specifications
57448 | |
purified | |
Human BRUCE | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q9NR09 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
BIRC6 | |
0.02% sodium azide | |
50 μg | |
PBS |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.