Learn More
Thermo Scientific™ HyperSep™ SCX Strong Cation Exchanger SPE 96-Well Plates and Individual Wells
Obtain excellent performance for extraction of charged basic compounds with these strong cation exchanger SPE well plates and individual wells.
Brand: Thermo Scientific™ 60300-582
2746.68 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Thermo Scientific™ HyperSep SCX Strong Cation Exchanger SPE 96-Well Plates and Individual Wells feature a strong cation exchange sorbent for extraction of charged basic compounds. Typical applications include extraction of:
Antibiotics
Drug compounds
Organic bases
Amino acids
Catecholamines
Herbicides
HyperSep SCX Strong Cation Exchanger 96-Well Plates and Individual Wells are ideally suited for the extraction of a wide range of charged basic compounds.Modified benzene ring provides potential for nonpolar interactions
Low pKa value
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
Specifications
Individual SPE wells; Bed weight 25 mg | |
40 to 60 μm | |
HyperSep SCX |
Extraction of Charged Basic Compounds | |
HyperSep | |
Individual Well |