Learn More
Idoxuridine, MedChemExpress
MedChemExpress Idoxuridine (5-Iodo-2′-deoxyuridine, 5-IUdR, IdUrd) is an iodinated thymidine analogue that competitively inhibits phosphorylases. Idoxuridine can inhibit viral activity, particularly viral eye infections, including herpes simplex keratitis, by inhibiting DNA polymerase and affecting viral replication. Idoxuridine against feline herpesvirus has the IC50 value of 4.3 μM. Idoxuridine shows anti-orthopoxvirus activity.
Brand:  Medchem Express HY-B0307
Encompass
 View more versions of this product
Product Code. 16418448
  • POA / 10g
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
Idoxuridine (5-Iodo-2′-deoxyuridine, 5-IUdR, IdUrd) is an iodinated thymidine analogue that competitively inhibits phosphorylases. Idoxuridine can inhibit viral activity, particularly viral eye infections, including herpes simplex keratitis, by inhibiting DNA polymerase and affecting viral replication. Idoxuridine against feline herpesvirus has the IC50 value of 4.3 μM. Idoxuridine shows anti-orthopoxvirus activity.
Specifications
Specifications
Idoxuridine
White
C9H11IN2O5
5-Iodo-2′-deoxyuridine 5-IUdR IdUrd
OC[C@@H]1[C@H](C[C@H](N2C(NC(C(I)=C2)=O)=O)O1)O
354.1
Research
COVID-19-anti-virus
54-42-2
Research
10 g
DMSO : 125 mg/mL (353.01 mM; Need ultrasonic)
354.1
98.0%
Powder
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.