missing translation for 'onlineSavingsMsg'
Learn More

Idoxuridine, MedChemExpress

MedChemExpress Idoxuridine (5-Iodo-2′-deoxyuridine, 5-IUdR, IdUrd) is an iodinated thymidine analogue that competitively inhibits phosphorylases. Idoxuridine can inhibit viral activity, particularly viral eye infections, including herpes simplex keratitis, by inhibiting DNA polymerase and affecting viral replication. Idoxuridine against feline herpesvirus has the IC50 value of 4.3 μM. Idoxuridine shows anti-orthopoxvirus activity.

POA - 2143.91 DKK

Specifications

Chemical Name or Material Idoxuridine
CAS 54-42-2
Color White
Purity Grade Notes Research
Molecular Formula C9H11IN2O5
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16470868
Encompass_Preferred
View Documents
Medchem Express
HY-B0307
5 g
2143.91 DKK
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16462129
Encompass_Preferred
View Documents
Medchem Express
HY-B0307
1 g
714.64 DKK
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16418448
Encompass
View Documents
Medchem Express
HY-B0307
10 g
POA
10g
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

Idoxuridine (5-Iodo-2′-deoxyuridine, 5-IUdR, IdUrd) is an iodinated thymidine analogue that competitively inhibits phosphorylases. Idoxuridine can inhibit viral activity, particularly viral eye infections, including herpes simplex keratitis, by inhibiting DNA polymerase and affecting viral replication. Idoxuridine against feline herpesvirus has the IC50 value of 4.3 μM. Idoxuridine shows anti-orthopoxvirus activity.
Specifications

Specifications

Idoxuridine
White
C9H11IN2O5
DMSO : 125 mg/mL (353.01 mM; Need ultrasonic)
354.1
98.0%
Powder
54-42-2
Research
5-Iodo-2′-deoxyuridine 5-IUdR IdUrd
OC[C@@H]1[C@H](C[C@H](N2C(NC(C(I)=C2)=O)=O)O1)O
354.1
Research
COVID-19-anti-virus
Videos
Safety and Handling

Safety and Handling

H302∣H315∣H319∣H335

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.