Learn More
Idoxuridine, MedChemExpress
MedChemExpress Idoxuridine (5-Iodo-2′-deoxyuridine, 5-IUdR, IdUrd) is an iodinated thymidine analogue that competitively inhibits phosphorylases. Idoxuridine can inhibit viral activity, particularly viral eye infections, including herpes simplex keratitis, by inhibiting DNA polymerase and affecting viral replication. Idoxuridine against feline herpesvirus has the IC50 value of 4.3 μM. Idoxuridine shows anti-orthopoxvirus activity.
Brand: Medchem Express HY-B0307
Description
Idoxuridine (5-Iodo-2′-deoxyuridine, 5-IUdR, IdUrd) is an iodinated thymidine analogue that competitively inhibits phosphorylases. Idoxuridine can inhibit viral activity, particularly viral eye infections, including herpes simplex keratitis, by inhibiting DNA polymerase and affecting viral replication. Idoxuridine against feline herpesvirus has the IC50 value of 4.3 μM. Idoxuridine shows anti-orthopoxvirus activity.Specifications
Idoxuridine | |
White | |
C9H11IN2O5 | |
5-Iodo-2′-deoxyuridine 5-IUdR IdUrd | |
OC[C@@H]1[C@H](C[C@H](N2C(NC(C(I)=C2)=O)=O)O1)O | |
354.1 | |
Research | |
COVID-19-anti-virus |
54-42-2 | |
Research | |
10 g | |
DMSO : 125 mg/mL (353.01 mM; Need ultrasonic) | |
354.1 | |
98.0% | |
Powder |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only