Learn More
BINDER™ Incubator Base
Incubator Base equipped with casters is used for the secure positioning and leveling of a BINDER™ CO2 incubator.
Brand:  BINDER™ 9051-0044
Product Code. 16684183
  • 4105.00 DKK / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • For the secure positioning and leveling of a BINDER™ CO2 incubator
  • Suitable for:
    • CB 260
    • CB-S 260
Specifications
Specifications
Base - the base equipped with casters is used for the secure positioning and leveling of a BINDER CO2 incubator
Steel
Casters
18.74 lb.
BINDER Chambers Chambers CB260, CB-S260, CBF 260
Base
920 x 920 x 350 mm
8.5 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BINDER™ Incubator Base > For Use With: BINDER Units CB260 and CB-S260