Learn More
Memmert™ Interior Socket
Interior IP68 socket (for temperature restriction +70°C option) 230V, 2. A. Cannot be switched individually, moisture tight IP68
Brand:  Memmert™ HPP410ecoR3
 View more versions of this product
Product Code. 17129844
  • 3665.00 DKK / Each
Estimated Shipment: 26-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Accessory
  • Ampacity 230 V/2.2 A
  • Can be switched off with the On/Off switch
Specifications
Specifications
HPP410eco
230 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Interior Socket > HPP410eco