Learn More
Cytiva INV-907 ETFE Titanium Fill Port
Used for small samples. Cytiva INV-907 ETFE Titanium Fill Port is designed for biocompatibillity.
Brand: Cytiva 18-1127-66
1586.50 DKK valid until 2025-05-31
Use promo code "24127" to get your promotional price.
Description
- A range of manual and automated valves to meet a variety of performance requirements.
- Designed for biocompatibility and years of reliable service, each may be inserted into the flow path without causing unnecessary mixing or sample dilution.
Specifications
Fill Port | |
ETFE, Titanium | |
For Use with All Aqueous Solutions and Organic Solvents Commonly Employed in Chromatography |
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
26Hours
4Minutes
18Seconds
43