Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - CO2 and Temperature
For one free-selectable CO2 and temperature value
Brand: Memmert™ ICO150D38898
This item is not returnable.
View return policy
Description
- Includes temperature distribution survey at Memmert
- For 27 measuring points to DIN 12880:2007-05
- PQ check list as support for validation by customer.
Specifications
ICO150 | |
IQ/OQ Document with Device-specific Works Test Data |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - CO2 and Temperature > ICO150
Spot an opportunity for improvement?Share a Content Correction