Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - CO2 and Temperature
For one free-selectable CO2 and temperature value
17320.00 DKK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
17109964
|
Memmert™
ICO150D38898 |
ICO150 | N/A | N/A | N/A | |||||
17149994
|
Memmert™
ICO240D38898 |
ICO240 | N/A | N/A | N/A | |||||
17170025
|
Memmert™
ICO50D38898 |
ICO50 | N/A | N/A | N/A | |||||
17179924
|
Memmert™
ICO105D38898 |
ICO105 | N/A | N/A | N/A | |||||
Description
- Includes temperature distribution survey at Memmert
- For 27 measuring points to DIN 12880:2007-05
- PQ check list as support for validation by customer.
Specifications
IQ/OQ Document with Device-specific Works Test Data |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - CO2 and Temperature