Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - Temperature
For one free-selectable temperature value, Includestemperature distribution survey at Memmert to DIN 12880:2007-05
Brand: Memmert™ HPP1060ecoD00127
This item is not returnable.
View return policy
Description
- 27 measuring points (sizes 110 - 1060, 2200)
- 26 measuring points (size 1400)
- PQ checklist as support for validation by customer
Specifications
HPP1060eco | |
IQ/OQ Document with Device-specific Works Test Data |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - Temperature > HPP1060eco
Spot an opportunity for improvement?Share a Content Correction