Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - Temperature
For one free-selectable temperature value, Includestemperature distribution survey at Memmert to DIN 12880:2007-05
11120.00 DKK - 16185.00 DKK
Specifications
Product Type IQ/OQ Document with Device-specific Works Test Data
View More Specs
Products 13
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
17100135
View Documents
Memmert™
IPP1400ecoD00127
IPP1400eco
16185.00 DKK
Each
17100185
View Documents
Memmert™
IPP260ecoD00127
IPP260eco
16185.00 DKK
Each
17109684
View Documents
Memmert™
HPP110ecoD00127
HPP110eco This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17110155
View Documents
Memmert™
IPP2200ecoD00127
IPP2200eco
16185.00 DKK
Each
17110265
View Documents
Memmert™
IPP750ecoD00127
IPP750eco
16185.00 DKK
Each
17129744
View Documents
Memmert™
HPP2200ecoD00127
HPP2200eco This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17149774
View Documents
Memmert™
HPP260ecoD00127
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17170055
View Documents
Memmert™
IPP1060ecoD00127
IPP1060eco
16185.00 DKK
Each
17179864
View Documents
Memmert™
HPP750ecoD00127
HPP750eco This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17189714
View Documents
Memmert™
HPP1400ecoD00127
HPP1400eco This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17190215
View Documents
Memmert™
IPP410ecoD00127
IPP410eco
16185.00 DKK
Each
17199634
View Documents
Memmert™
HPP1060ecoD00127
HPP1060eco This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17199814
View Documents
Memmert™
HPP410ecoD00127
HPP410eco This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • 27 measuring points (sizes 110 - 1060, 2200)
  • 26 measuring points (size 1400)
  • PQ checklist as support for validation by customer
Specifications
Specifications
IQ/OQ Document with Device-specific Works Test Data
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - Temperature