Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data- Temperature and Humidity
For one free-selectable temperature and humidity value, and measuring of light intensity
Brand: Memmert™ HPP260ecoD00137
This item is not returnable.
View return policy
Description
- Includes temperature distribution survey at Memmert
- For 27 measuring points to DIN 12880:2007-05
- PQ check list as support for validation by customer.
Specifications
HPP260eco | |
IQ/OQ Document with Device-specific Works Test Data |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ IQ/OQ Document with Device-Specific Works Test Data- Temperature and Humidity > HPP260eco
Spot an opportunity for improvement?Share a Content Correction